Mercury(I) nitrate dihydrate, ACS reagent, 9th edition, Thermo Scientific Chemicals
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(I) nitrate dihydrate, ACS reagent, 9th edition, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447182500
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
Add to cart
Mercury(I) nitrate dihydrate, ACS reagent, 9th edition, Thermo Scientific Chemicals
Catalog number447182500
Price (USD)/ Each
292.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
344.00 
Save 51.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemercury(1+) hydrate nitrate
Molecular FormulaH2HgNO4
InChI KeyUPCSQZXLTBTBCU-UHFFFAOYSA-N
SMILESO.[Hg+].[O-][N+]([O-])=O
Molecular Weight (g/mol)280.61
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after reduction=<0.01 %
Chloride (Cl)=<0.005 %
Sulfate (SO4)=<0.005 %
Iron (Fe)=<0.001 %
Mercuric Mercury (as Hg)=<0.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only