Thermo Scientific Chemicals

Potassium chromate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447205000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Potassium chromate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447205000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 447205000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7789-00-6
IUPAC Name
dipotassium dioxochromiumbis(olate)
Molecular Formula
CrK2O4
InChI Key
XMXNVYPJWBTAHN-UHFFFAOYSA-N
SMILES
[K+].[K+].[O-][Cr]([O-])(=O)=O
pH
8.6 to 9.8 (5% soln. at 25°C)
Insoluble matter
=<0.005 %
Chloride (Cl)
=<0.005 %
Sulfate (SO4)
=<0.03 %
Calcium (Ca)
=<0.005 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text