Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™
Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™
Thermo Scientific Chemicals

Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroSeal™

CAS: 1330-20-7 | C8H10 | 106.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4472400101 LAcroSeal™ Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447240010
Price (USD)
-
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Lead (Pb)=<0.2 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Share catalog number, name or link