Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals
Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 447261000
Price (USD)/ Each
56.65
Online exclusive
62.40 
Save 5.75 (9%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
62.40 
Save 5.75 (9%)
Add to cart
Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals
Catalog number447261000
Price (USD)/ Each
56.65
Online exclusive
62.40 
Save 5.75 (9%)
-
Add to cart
Chemical Identifiers
CAS7803-55-6
IUPAC Namevanadiumoylol amine
Molecular FormulaH4NO3V
InChI KeyYBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILESN.O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO4>=99.0 %
Solubility(in Ammonium hydroxide) Passes test
Carbonate (CO3)(Limit about 0.3 %) Passes test
Chloride (Cl)=<0.2 %
Appearance (Color)White to yellow to tan
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only