Thermo Scientific Chemicals

Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447265000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium metavanadate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447265000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 447265000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7803-55-6
IUPAC Name
vanadiumoylol amine
Molecular Formula
H4NO3V
InChI Key
YBVKNHXQSRDWAA-UHFFFAOYSA-M
SMILES
N.O[V](=O)=O
Titration with KMnO4
>=99.0 %
Solubility
(in Ammonium hydroxide) Passes test
Carbonate (CO3)
(Limit about 0.3 %) Passes test
Chloride (Cl)
=<0.2 %
Appearance (Color)
White to yellow to tan

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text