Mercuric acetate, ACS reagent, Thermo Scientific Chemicals
Mercuric acetate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercuric acetate, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447271000
Price (USD)/ Each
120.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
142.00 
Save 21.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
120.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
142.00 
Save 21.35 (15%)
Add to cart
Mercuric acetate, ACS reagent, Thermo Scientific Chemicals
Catalog number447271000
Price (USD)/ Each
120.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
142.00 
Save 21.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Titration with NH4SCN>=98.0 %
Insoluble matter=<0.01 %
Nitrate (NO3)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only