These are replacement tip combs for the Pharmaceutical Analytics KingFisher™ Flex 96 PCR Head Magnetic Particle Processor and MagMAX™ Express-96 Deep Well Magnetic Particle Processor with PCR Head.
Optimize performance using MagMax/KingFisher system consumables designed specifically for use with these instruments. MagMax/KingFisher instruments utilize disposable plastics made of polypropylene. The plastics—tip combs and microplates—are ideal for magnetic particle processing because of their low binding affinity for biomolecules. The special design enables excellent recovery of magnetic beads.
Contains eighty (80) pieces dual-packaged in ten (10) paperboard boxes packed into one (1) cardboard box.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format
96-well Plate
Capacity
96 Samples
Product Line
MagMAX™
Shipping Condition
Room Temperature
Contents & Storage
Contains eighty (80) pieces dual-packaged in ten (10) paperboard boxes packed into one (1) cardboard box.