Sodium hydroxide, pure, pellets, Thermo Scientific Chemicals
Sodium hydroxide, pure, pellets, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hydroxide, pure, pellets, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 447280050
Price (USD)/ Each
73.65
Online exclusive
81.60 
Save 7.95 (10%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
81.60 
Save 7.95 (10%)
Add to cart
Sodium hydroxide, pure, pellets, Thermo Scientific Chemicals
Catalog number447280050
Price (USD)/ Each
73.65
Online exclusive
81.60 
Save 7.95 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2
IUPAC Namesodium hydroxide
Molecular FormulaHNaO
InChI KeyHEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES[OH-].[Na+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pellets
Sodium carbonate (Na2CO3)=<2.0 %
Iron (Fe)=<0.001 %
Appearance (Color)White to almost white
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only