Thermo Scientific Chemicals

Citric acid, trisodium salt dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 447290010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Citric acid, trisodium salt dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 447290010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 447290010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6132-04-3
IUPAC Name
trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular Formula
C6H9Na3O9
InChI Key
NLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILES
O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
White
pH
7.5 to 9.0
UV
at 260 nm A: =<0.01
Appearance (Form)
Crystalline powder
UV
(0.1 M in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text