Sodium chloride, 99.5%, for biochemistry
Sodium chloride, 99.5%, for biochemistry
Thermo Scientific Chemicals

Sodium chloride, 99.5%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number 447300010
Price (USD)/ Each
71.65
Online exclusive
83.60 
Save 11.95 (14%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
83.60 
Save 11.95 (14%)
Add to cart
Sodium chloride, 99.5%, for biochemistry
Catalog number447300010
Price (USD)/ Each
71.65
Online exclusive
83.60 
Save 11.95 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
Molecular Weight (g/mol)58.44
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.4 %
UVat 260 nm A: =<0.01
Appearance (Form)Powder or crystals
UV(0.1 M in water)
UVat 280 nm A: =<0.01
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only