Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 447310010
Price (EUR)
232,05
Online Exclusive
280,00
Save 47,95 (17%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
232,05
Online Exclusive
280,00
Save 47,95 (17%)
Each
Add to cart
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.005 %
pH3.8 to 4.5 (1 M in water)
SolubilityClear colorless
UVat 280 nm A: =<0.01
Heavy metals (as Pb)=<10 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Safety and Handling

Not Hazardous

Not Hazardous

Label Elements

Share catalog number, name or link