Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 99.5%, for biochemistry

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 447312500
Price (USD)
70.65
Special offer
Online exclusive
Ends: 26-Sep-2025
86.70
Save 16.05 (19%)
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 26-Sep-2025
86.70
Save 16.05 (19%)
Each
Add to cart
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.005 %
pH3.8 to 4.5 (1 M in water)
SolubilityClear colorless
UVat 280 nm A: =<0.01
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link