Thermo Scientific Chemicals

Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 447320025
2.5 kg, Plastic Bottle, Each
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Catalog number: 447320025
2.5 kg, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 447320025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10035-04-8
IUPAC Name
calcium dihydrate dichloride
Molecular Formula
CaCl2H4O2
InChI Key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ca++]
Appearance (Form)
Crystalline powder
Heavy metals (as Pb)
=<10 ppm
Alkalinity
Passes test
Solubility
Clear colorless
Acidity
Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text