Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals
Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 447325000
Price (USD)/ Each
56.65
Online exclusive
66.40 
Save 9.75 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
66.40 
Save 9.75 (15%)
Add to cart
Calcium chloride dihydrate, 99%, for biochemistry, Thermo Scientific Chemicals
Catalog number447325000
Price (USD)/ Each
56.65
Online exclusive
66.40 
Save 9.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium dihydrate dichloride
Molecular FormulaCaCl2H4O2
InChI KeyLLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Ca++]
Molecular Weight (g/mol)147.01
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric>=98.5 %
Aluminium (Al)=<1 ppm
SolubilityClear colorless
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only