Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447405000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Molybdenum(VI) oxide, 99.5+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447405000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 447405000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1313-27-5
IUPAC Name
trioxomolybdenum
Molecular Formula
MoO3
InChI Key
JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES
O=[Mo](=O)=O
Appearance (Form)
Powder
Ammonium (NH4)
=<0.002 %
Titration
>=99.5 % (Precipitation titration of molybdate)
Chloride (Cl)
=<0.002 %
Phosphate (PO4)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text