Thermo Scientific Chemicals

1,2-Propanediol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447410025
2.5 L, Each
Thermo Scientific Chemicals

1,2-Propanediol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447410025
2.5 L, Each
Quantity
Catalog number: 447410025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-55-6
IUPAC Name
propane-1,2-diol
Molecular Formula
C3H8O2
InChI Key
DNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES
CC(O)CO
Residue after ignition
=<0.005 %
Water
=<0.2 %
Chloride (Cl)
=<1 ppm
Color scale
=<10 APHA
Appearance (Form)
Clear viscous liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text