1x1 image pixel for data collection
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
1,2-Propanediol, ACS reagent
Thermo Scientific Chemicals

1,2-Propanediol, ACS reagent

CAS: 57-55-6 | C3H8O2 | 76.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Catalog number 447411000
Price (USD)
36.65
Online Exclusive
40.60
Save 3.95 (10%)
Each
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
40.60
Save 3.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS57-55-6
IUPAC Namepropane-1,2-diol
Molecular FormulaC3H8O2
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILESCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Chloride (Cl)=<1 ppm
Residue after ignition=<0.005 %
Color scale=<10 APHA
Appearance (Form)Clear viscous liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link