Informational:Need help deciding what product to use with the Ion PGM™ System? Check out our helpful Applications Decision Tree for the Ion PGM™ System.
Ion Torrent™

Ion Xpress™ Barcode Adapters 49-64 Kit

Catalog number: 4474519
Have Questions?
Ion Torrent™

Ion Xpress™ Barcode Adapters 49-64 Kit

Catalog number: 4474519
Have Questions?
Catalog Number
4474519
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
44745191 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Xpress™ Barcode Adapters 49-64 Kit provides a set of 16 unique barcode adaptors specifically designed and validated for optimal performance with the Ion Torrent PGM™ and Proton™ semiconductor sequencers. When used in combination with the Ion Xpress™ Plus Fragment Library Kit or the Ion Plus Fragment Library Kit, this kit enables users to pool up to 16 fragment libraries prior to emulsion PCR and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for a wide range of applications, including targeted enrichment. Use of this kit with other Barcode Adapters kits allows pooling of up to 96 fragment libraries.

Key Product Features:

• Enables multiplexing of numerous fragment library samples on a single sequencing chip with the use of robust molecular barcodes
• Both sequence- and flow-optimized for equal representation all barcodes in a pool and more economical multiplexed sequencing runs
• Robust error correction built in by design for added confidence in sample identification

Multiplexing with Barcode Adaptors Enables Higher Throughput
Multiplexing with barcoded libraries leads to more cost effective runs by permitting up to 96 samples per run (16 with this kit), significantly decreasing the cost and handling requirements of emPCR, enrichment, and sequencing.

Sequence- and Flow-Optimized for Increased Performance and Efficiency
Barcode adapters contain sequences that are optimized to provide equal representation of all barcodes in a pool, require a minimum number of flows to interrogate, and require only 17 bases of adaptor sequencing required which results in more economical multiplexed sequencing runs.

Robust Error Correction Built In by Design
Built-in error correction virtually eliminates the possibility of incorrect sample assignment.

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line
Ion Torrent™
Starting Material
DNA
For Use With (Equipment)
Ion PGM™ System, Ion OneTouch™ System
Libraries
Fragment Library
Product Type
Barcode Adapter Kit
Sequencing Type
Genome & DNA Sequencing
Shipping Condition
Dry Ice
Throughput
96 Samples per Run
Workflow Step
Library Barcoding

Contents & Storage

1 box of 17 tubes; store -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text