Applied Biosystems™
SeqScape™ Software v3.0, Initial License
Catalog number: 4474978
Have Questions?
Applied Biosystems™
SeqScape™ Software v3.0, Initial License
Catalog number: 4474978
Have Questions?
Catalog Number
4474978
Unit Size
1 license
Price (USD)
Catalog NumberUnit SizePrice (USD)
44749781 license
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This is an Initial License for the SeqScape™ Software. The SeqScape™ Software is a resequencing package designed for mutation detection and analysis, SNP discovery and validation, pathogen sub-typing, allele identification, and sequence confirmation. It provides library functions for comparison to a known group of sequences, as well as 21 CFR Part 11 functionalities.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Line
SeqScape™
Software Type
Sequencing Data Analysis
Data Entry
CE Sequencing (.ab1 files)
License
Initial
Operating System
Windows 7 Pro
Product Type
SeqScape Software V3.0 Initial License
Software Category
Supports files from all AB Genetic Analyzers
Contents & Storage
Software/CD