Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 447530010
1 kg, Each
Thermo Scientific Chemicals

Calcium nitrate tetrahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 447530010
1 kg, Each
Quantity
Catalog number: 447530010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13477-34-4
IUPAC Name
calcium bis(nitric acid) tetrahydrate
Molecular Formula
CaH10N2O10
InChI Key
DKCJOKYHYIAZQH-UHFFFAOYSA-N
SMILES
O.O.O.O.[Ca++].O[N+]([O-])=O.O[N+]([O-])=O
Titration Complexometric
97.5 to 102.5 %
Solubility
(20 g + 100 ml water) Clear colorless solution
Appearance (Form)
Crystals
pH
5.0 to 9.0 (5 % at 25°C)
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text