Solubility(20 g + 100 ml water) Clear colorless solution
Appearance (Form)Crystals
pH5.0 to 9.0 (5 % at 25°C)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.