Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 447540050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, extra pure, Thermo Scientific Chemicals

Catalog number: 447540050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 447540050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13446-18-9
IUPAC Name
magnesium(2+) hexahydrate dinitrate
Molecular Formula
H12MgN2O12
InChI Key
MFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
White
Titration Complexometric
98 to 102 %
Heavy metals (as Pb)
=<0.001 %
Appearance (Form)
Crystals
pH
5 to 8.2 (5 % in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text