DynaLight™ Substrate with RapidGlow™ Enhancer
DynaLight™ Substrate with RapidGlow™ Enhancer
Invitrogen™

DynaLight™ Substrate with RapidGlow™ Enhancer

Achieve faster ELISA results with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44754101 L
4475406100 mL
2 Options
Catalog number 4475410
Price (USD)/ 1 L
5,220.00
-
Add to cart
Quantity:
1 L
Price (USD)/ 1 L
5,220.00
Add to cart
DynaLight™ Substrate with RapidGlow™ Enhancer
Catalog number4475410
Price (USD)/ 1 L
5,220.00
-
Add to cart
Achieve faster ELISA results with DynaLight™ Substrate with RapidGlow™ Enhancer, a ready-to-use chemiluminescent substrate formulation that has been optimized to achieve faster results in solution-based assays. The DynaLight™ Substrate with RapidGlow™ Enhancer formulation includes 1,2-dioxetane chemiluminescent substrate and a polymeric enhancer that enables ultra-sensitive immunoassay detection by alkaline phosphatase label.

Features of DynaLight™ Substrate with RapidGlow™ Enhancer:

• Faster kinetics and higher signal levels than colorimetric, fluorescent, or other chemiluminescent chemistries
• Sensitive attomole detection of purified alkaline phosphatase enzyme
• Optimized formulation for both microplate and bead-based assays
• Substrate stability of over 1 year at 4°C
• Compatibility with automated platforms with trigger solution/positions already in place

The DynaLight™ Substrate with RapidGlow™ Enhancer provides a robust chemiluminescent reaction when catalyzed by alkaline phosphatase. This reaction achieves maximum light emission within 2 minutes at 37°C and maintains a sustained signal for hours. When combined with DynaLight™ Trigger Solution, DynaLight™ Substrate with RapidGlow™ Enhancer delivers maximum signal within seconds. The environmentally-friendly DynaLight™ Trigger Solution provides maximum signal within 1 second of addition, with a longer half-life to its ground state than other chemiluminescent Flash technologies, allowing ample time to collect data reliably.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
Product TypeELISA Substrate
Quantity1 L
Detection MethodChemiluminescence
Assay Sensitivity1 pg/mL
FormLiquid
FormatBottle
For Use With (Equipment)Luminometer (Microplate)
High-throughput CompatibilityHigh-throughput Compatible
Product LineDynaLight™, NovaBright™, RapidGlow™
Sample TypeCell Cultures, Whole Blood, Serum
Substrate PropertiesChemical Substrate
Substrate TypeAlkaline Phosphatase
Test Time2 min.
Shipping ConditionRoom Temperature
Unit Size1 L
Contents & Storage
1 amber bottle — Store at 2 to 8°C