Thermo Scientific Chemicals

Lithium sulfate, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 447552500
250 g, Each
Thermo Scientific Chemicals

Lithium sulfate, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 447552500
250 g, Each
Quantity
Catalog number: 447552500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10377-48-7
IUPAC Name
dilithium(1+) sulfate
Molecular Formula
Li2O4S
InChI Key
INHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES
[Li+].[Li+].[O-]S([O-])(=O)=O
Potassium (K)
=<0.02 %
Iron (Fe)
=<0.005 %
Magnesium (Mg)
=<0.02 %
Appearance (Form)
Crystalline powder
Water
=<1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text