Thermo Scientific Chemicals

Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447585000
500 g, Each
Thermo Scientific Chemicals

Gallic acid monohydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447585000
500 g, Each
Quantity
Catalog number: 447585000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5995-86-8
IUPAC Name
3,4,5-trihydroxybenzoate
Molecular Formula
C7H5O5
InChI Key
LNTHITQWFMADLM-UHFFFAOYSA-M
SMILES
OC1=CC(=CC(O)=C1O)C([O-])=O
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=98.0 %
Insoluble matter
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text