Thermo Scientific Chemicals

Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447591000
100 g, Each
Thermo Scientific Chemicals

Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447591000
100 g, Each
Quantity
Catalog number: 447591000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10112-91-1
IUPAC Name
dimercuranebis(ylium) dichloride
Molecular Formula
Cl2Hg2
InChI Key
ZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES
[Cl-].[Cl-].[Hg+][Hg+]
Appearance (Color)
White to off-white
Appearance (Form)
Crystalline powder
Titration Iodometric
>=99.5 %
Residue after reduction
=<0.02 %
Mercuric Chloride
=<0.01 % (HgCl2)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text