Informational:Need help deciding what product to use with the Ion PGM™ System? Check out our helpful Applications Decision Tree for the Ion PGM™ System.
Informational:This product is no longer recommended for small RNA applications. Please contact your Field Applications Specialist or Technical Support for additional information.
Ion Torrent™

Ion Total RNA-Seq Kit v2

Catalog number: 4475936
Have Questions?
Ion Torrent™

Ion Total RNA-Seq Kit v2

Catalog number: 4475936
Have Questions?
Catalog Number
4475936
Unit Size
12 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
48 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
447593612 reactions
-
447978948 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ion Total RNA-Seq Kit v2 includes the reagents needed to prepare representative cDNA libraries for strand-specific RNA sequencing on the Ion GeneStudio S5, Ion Proton, or the Ion Personal Genome Machine (PGM) systems. The Ion GeneStudio S5 and Ion Proton systems are ideally suited for sequencing the whole transcriptome (ribosomal RNA depleted or polyA), while the Ion GeneStudio S5 and Ion PGM systems are suitable for viral and bacterial transcriptomes. Version 2 of the Ion Total RNA-Seq Kit is an improvement over the first generation kit. Magnetic bead-based purification replaces all of the filter cleanup steps and the total reaction time has been reduced to 6 hours.

Additional Features of the New Ion Total RNA-Seq Kit v2:

• Greater accuracy—SuperScript VILO and Platinum PCR SuperMix High Fidelity added for highest template fidelity
• Barcode compatible—works with Ion Xpress RNA-Seq Barcode 01-16 Kit for multiplexing
• Automation friendly—magnetic bead-based purification simplifies automation of library construction

As with the previous kit, the Ion Total RNA-Seq Kit v2:

• Preserves strand information—all mapped reads are aligned in the direction of transcription relative to the chromosomal strand
• Allows you to analyze different types of RNA—supports rRNA depleted total RNA, and poly(A) RNA

The Ion Total RNA-Seq Kit v2 is designed to make cDNA library preparation fast and flexible. It can be used to generate a representative cDNA library, flanked by specific sequences necessary for sequencing, from any type of RNA sample.

Designed as a complete solution with a common workflow, the Ion Total RNA-Seq Kit v2 combines optimized reagents and protocols for discovery of coding RNA, noncoding RNA, and alternative splice variants.

Whole transcriptome analysis
The whole transcriptome protocol enables construction of strand-specific libraries in approximately 5 hours. Starting with as little as 100 ng of total RNA, construct a library from 1 ng of poly(A) RNA or 25 ng of rRNA-depleted RNA following the RNA enrichment and library generation protocols provided in the manual. Because the libraries are not limited to cDNA derived only from poly(A) RNA, Ion Total RNA-Seq Kit libraries support a more thorough investigation of transcriptome complexity, capable of characterizing known and undocumented transcripts, including alternative splice variants, fusion transcripts, and SNPs.

Preserve strand information
Unlike methods that ligate adapters to double-stranded cDNA, the Ion Total RNA-Seq Kit v2 utilizes proprietary Ambion technology to attach the adapters in a directional manner that preserves strand information in the resulting libraries. In addition, both the 3' and 5' adapters are attached simultaneously, reducing ligation and clean-up steps.

Preserving strand orientation during library construction helps enable more accurate determination of the structure and expression level of transcripts, and can aid in the discovery of novel transcription regions from both the positive and negative genomic strands.

The Ion Total RNA-Seq Kit v2 is designed to create RNA libraries from up to 48 samples for whole transcriptome sequencing on the Ion GeneStudio S5, Ion Proton, or Ion PGM systems.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Sample Type
RNA
For Use With (Equipment)
Ion GeneStudio™ S5, Ion PGM™, Ion Proton™ Systems
No. of Reactions
12 or 48
Libraries
cDNA Library
Product Line
Ion Torrent™
Sequencing Type
Transcriptome Sequencing
Starting Material Amount
For RNA (rRNA-depleted): 1-100 ng mRNA
For whole transcriptome: ≥100 ng total RNA
Workflow Step
NGS Library Preparation

Contents & Storage

Ion RNA-Seq Core Kit v2

Store at -20°C to room temperature:

2 x 1.75 mL Nuclease-free Water

Store at -20°C:
1 x 20 μL 10X RNase III Reaction Buffer
1 x 20 μL RNase III
1 x 40 μL Hybridization Solution
1 x 150 μL 2X Ligation Buffer
1 x 30 μL Ligation Enzyme Mix
1 x 30 μL 2.5 mM dNTP Mix
1 x 56 μL 10X SuperScript™ III Enzyme Mix
1 x 900 μL Platinum™ PCR SuperMix High Fidelity
1 x 50 μL WT Control RNA (1 μg/μL HeLa total RNA)
1 x 10 μL Small RNA Control (1 μg/μL human placenta total RNA)
1 x 10 μL Small RNA Control (1 μg/μL human placenta total RNA)

Ion RNA-Seq Primer Set v2 (Cat. No. 4474810)

Store at -20°C:

1 x 30 μL Ion Adaptor Mix v2
1 x 104 μL Ion RT Primer v2
1 x 20 μL Ion 5' PCR Primer v2
1 x 20 μL Ion 3' PCR Primer v2

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text