Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447595000
Price (USD)/ Each
318.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
375.00 
Save 56.35 (15%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
318.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
375.00 
Save 56.35 (15%)
Add to cart
Mercury(I) chloride, ACS reagent, Thermo Scientific Chemicals
Catalog number447595000
Price (USD)/ Each
318.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
375.00 
Save 56.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10112-91-1
IUPAC Namedimercuranebis(ylium) dichloride
Molecular FormulaCl2Hg2
InChI KeyZOMNIUBKTOKEHS-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg+][Hg+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Titration Iodometric>=99.5 %
Residue after reduction=<0.02 %
Mercuric Chloride=<0.01 % (HgCl2)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only