Thermo Scientific Chemicals

Xylenol orange, sodium salt, ACS reagent

Catalog number: 447600010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Xylenol orange, sodium salt, ACS reagent

Catalog number: 447600010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 447600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3618-43-7
IUPAC Name
tetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular Formula
C31H28N2Na4O13S
InChI Key
FXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES
[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
Appearance (Color)
Red-brown to dark brown
Appearance (Form)
Crystalline powder
Clarity of solution
Passes test
Suitability
(for Zn titration) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text