Thermo Scientific Chemicals

Potassium bisulfate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 447610100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Potassium bisulfate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 447610100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 447610100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7646-93-7
IUPAC Name
potassium hydrogen sulfate
Molecular Formula
HKO4S
InChI Key
CHKVPAROMQMJNQ-UHFFFAOYSA-M
SMILES
[K+].OS([O-])(=O)=O
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.5 %
Solubility
(10 % in water) Clear colorless
Chloride (Cl)
=<0.002 %
Iron (Fe)
=<0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text