Ion Torrent™

Ion Plus Fragment Library Adapters

產品號碼: 4476340
Ion Torrent™

Ion Plus Fragment Library Adapters

產品號碼: 4476340
產品號碼
4476340
包裝規格
10 reactions
產品號碼包裝規格價格 (TWD)
447634010 reactions與我們聯繫 ›
產品概覽
圖表
影片
建議
建議
文件
常見問題
引用資料與參考文獻
其他資訊
建議
The Ion Plus Fragment Library Adapters consist of standard fragment library adapter oligos for library construction of 10-20 DNA libraries (depending on input DNA type and amount) for semiconductor sequencing. While library-creation kits like the Ion Plus Fragment Library Kit and Ion Xpress Plus Fragment Library Kit include standard library adapters, the need for additional adapters can sometimes arise. For example, these Ion Plus Fragment Library Adapters are suitable for use with the AB Library Builder™ System to construct automated fragment libraries. Additionally, the Long Mate-Pair Library protocol for the Ion PGM™ Sequencer requires users to purchase standard adapter oligos – a task made easy with the introduction of our Ion Plus Fragment Library Adapters.

The Ion Plus Fragment Library Adapters offer you:

• Amplification primers for PCR amplification of standard Ion Plus fragment libraries
• The convenience of premixed and appropriately diluted adapter oligos for use in Ion fragment library construction protocols including automated library construction on the AB Library Builder™ System

For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

規格

Sample Type
DNA
Sequencing Type
Genome and DNA Sequencing
For Use With (Equipment)
Ion PGM™ System
For Use With (Application)
Sequencing
Libraries
Fragment Library
No. of Reactions
10
Product Line
Ion Torrent™
Product Type
Ion Plus Fragment Library Adapters
Shipping Condition
Dry Ice
Workflow Step
Library Generation

內容物與存放

4 tubes; store -20°C

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text