1x1 image pixel for data collection
GlobalFiler™ Express PCR Amplification Kit
GlobalFiler™ Express PCR Amplification Kit
Applied Biosystems™

GlobalFiler™ Express PCR Amplification Kit

The GlobalFiler Express PCR Amplification Kit is the first 6-dye, 24-locus STR kit that combines maximum compatibility with global databasingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4476609200 Reactions
44746651000 Reactions
Catalog number 4476609
Price (USD)
5,210.00
Each
-
Add to cart
No. of Reactions:
200 Reactions
Price (USD)
5,210.00
Each
Add to cart
Ask our AI about this Product
The GlobalFiler Express PCR Amplification Kit is the first 6-dye, 24-locus STR kit that combines maximum compatibility with global databasing loci standards, with dramatically reduced amplification time and superior discrimination power, to enable forensic DNA labs worldwide to maximize information recovery and improve overall efficiency. The GlobalFiler Express kit is part of a fully integrated and validated forensic DNA workflow backed by best-in-class global training, service, and support. This kit is optimized for database and single-source samples; for casework samples, please see our GlobalFiler PCR Amplification Kit.

Key Features:

• Up to 5-times faster amplification time than previous-generation kits
• DNA results in <2 hours—using current CE platforms
• Optimized protocols for blood and buccal samples deposited on swab or paper substrates
• The only kit with all markers recommended for inclusion by the CODIS Core Loci Working Group, including all markers commonly used in Europe, for maximum global database compatibility

The GlobalFiler Express kit is a robust and reliable single-amplification STR multiplex kit optimized for use with database and single-source samples. The kit leverages new dyes for the incorporation of additional markers, generating the most discriminating kit available to the forensic community. The kit contains all of the markers included in the major global databases and utilizes optimized reagents, thermal cycling parameters, and the latest advances in oligo synthesis and purification to enable the highest quality results specific to the sample type being processed. Maintenance of the NGM SElect and Identifiler primer sequences helps to ensure maximum concordance with previously typed samples. In addition, multiple gender markers have been added to the multiplex to enable accurate typing of male samples, even highly degraded or Y-negative samples, while increased genotyping accuracy is obtained with the expansion of the allelic ladder allele ranges to include all rare alleles as described in the literature (STRbase).

Experience Matters
We are the only company in the world that designs and validates its reagents, instruments, and data analysis software together as an integrated system for DNA testing. Technical service scientists and field application specialists are available to answer your technical questions, review data, and make recommendations on how to perform assays and troubleshoot results. All of these resources help ensure success when using Applied Biosystems human identification solutions in your laboratory.
Specifications
Product LineGlobalFiler™
Product TypePCR Amplification Kit
Quantity200 reactions
Shipping ConditionDry Ice
No. of Reactions200 Reactions
Starting MaterialDNA
Unit SizeEach
Contents & Storage
Store at -15°C to -25°C upon receipt and 2 to 8°C after initial use for up to 6 months.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link