Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 447670050
5 kg, Each
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 447670050
5 kg, Each
Quantity
Catalog number: 447670050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-77-0
IUPAC Name
potassium dihydrogen phosphate
Molecular Formula
H2KO4P
InChI Key
GNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES
[K+].OP(O)([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.5 %
Loss on drying
=<0.2 %
pH
4.2 to 4.5 (5 % solution)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text