Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals
Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
View all
Plastic bottle
Catalog number 447710010
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
Add to cart
Lithium acetate dihydrate, 98%, for analysis, Thermo Scientific Chemicals
Catalog number447710010
Price (USD)/ Each
155.65
Online exclusive
173.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS6108-17-4
IUPAC Namelithium(1+) acetate dihydrate
Molecular FormulaC2H7LiO4
InChI KeyIAQLJCYTGRMXMA-UHFFFAOYSA-M
SMILES[Li+].O.O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HClO497.5 to 102.5 %
Insoluble matter=<0.01 % (in water)
pH7.5 to 9.5 (5 % in water)
Chloride (Cl)=<0.003 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only