Thermo Scientific Chemicals

Benzyl alcohol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447721000
100 mL, Glass Bottle, Each
Thermo Scientific Chemicals

Benzyl alcohol, ACS reagent, Thermo Scientific Chemicals

Catalog number: 447721000
100 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 447721000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
100-51-6
IUPAC Name
phenylmethanol
Molecular Formula
C7H8O
InChI Key
WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES
OCC1=CC=CC=C1
Ash
=<0.005 %
Color scale
=<20 APHA
GC
>=99.0 %
GC
=<0.02 % Acetophenone
GC
=<0.01 % Benzaldehyde

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text