Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 447740050
5 kg, Each
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 447740050
5 kg, Each
Quantity
Catalog number: 447740050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10049-21-5
IUPAC Name
sodium phosphoric acid hydrate
Molecular Formula
H5NaO5P
InChI Key
BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES
O.[Na+].OP(O)(O)=O
Titration with NaOH
97.5 to 102.5 %
Insoluble matter in water
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text