Thermo Scientific Chemicals

Bismuth(III) bromide, 97%, Thermo Scientific Chemicals

Catalog number: 447771000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

Bismuth(III) bromide, 97%, Thermo Scientific Chemicals

Catalog number: 447771000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 447771000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-58-8
IUPAC Name
bismuth(3+) tribromide
Molecular Formula
BiBr3
InChI Key
TXKAQZRUJUNDHI-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Bi+3]
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
Light yellow to yellow or light grey
Assay
96 to 103 % (Bi)
Melting point
218°C to 220°C
Chloride (Cl)
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text