Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Sodium benzoate, 99%, for biochemistry
Thermo Scientific Chemicals

Sodium benzoate, 99%, for biochemistry

CAS: 532-32-1 | C7H5NaO2 | 144.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number 447800010
Price (USD)
88.65
Online Exclusive
98.10
Save 9.45 (10%)
Each
-
Add to cart
Quantity:
1 kg
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
88.65
Online Exclusive
98.10
Save 9.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS532-32-1
IUPAC Namesodium benzoate
Molecular FormulaC7H5NaO2
InChI KeyWXMKPNITSTVMEF-UHFFFAOYSA-M
SMILES[Na+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Titration with HClO4>=98.5 %
Loss on drying=<1 %
Solubility(1 M in water) Clear colorless to light yellow
Mercury (Hg)=<0.0001 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection