Thermo Scientific Chemicals

Potassium nitrate, 99+%, for biochemistry

Catalog number: 447820025
2.5kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium nitrate, 99+%, for biochemistry

Catalog number: 447820025
2.5kg, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 447820025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7757-79-1
IUPAC Name
potassium nitrate
Molecular Formula
KNO3
InChI Key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES
[K+].[O-][N+]([O-])=O
Sulfate (SO4)
=<0.015 %
Titration after Ion exchange
>=99.0 %
Heavy metals (as Pb)
=<10 ppm
Solubility
(1 M in water) Clear colorless
pH
4.5 to 8.5

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text