1x1 image pixel for data collection
Total Exosome Isolation Reagent (from serum)
Total Exosome Isolation Reagent (from serum)
Invitrogen™
Total Exosome Isolation Reagent (from serum)
Total Exosome Isolation Reagent (from serum) enables fast and efficient enrichment of intact exosomes from blood serum samples.• Maximise recoveryRead more
Have Questions?
Catalog NumberQuantity
44783606 mL
Catalog number 4478360
Price (USD)
481.00
Each
In stock
Add to cart
Quantity:
6 mL
Price (USD)
481.00
Each
Add to cart
Total Exosome Isolation Reagent (from serum) enables fast and efficient enrichment of intact exosomes from blood serum samples.
• Maximise recovery of intact exosomes for any type of downstream applications
• Easily isolate exosomes with our simple and reliable protocol
• Avoid time-consuming ultra-centrifugation
• Maximize flexibility—can be scaled up or down depending on the sample size
Isolation of exosomes can be a tedious, non-specific, and difficult process. Total Exosome Isolation Reagent (from serum) enables simple and reliable concentration of intact exosomes from blood serum samples with a protocol that is scalable depending on your sample size.
By tying up water molecules, the reagent forces less-soluble components such as vesicles out of solution, allowing them to be collected by a short, low-speed centrifugation. The reagent is added to the serum sample, and the solution is incubated at 2°C to 8°C for 30 min. The precipitated exosomes are recovered by standard centrifugation at 10,000 x g for 10 min. The pellet is then resuspended in PBS or similar buffer, and the exosomes are ready for downstream analysis or further purification by affinity methods. Total RNA and protein can then be purified using theTotal Exosome RNA and Protein Isolation Kit, or the intact exosomes can be used for biological studies of their pathways, functions, and trafficking. For immunoprecipitation of exosomal proteins, use Exosome Immunoprecipitation (Protein A) or Exosome Immunoprecipitation (Protein G).
To isolate intact exosomes from cell culture media, we recommend Total Exosome Isolation Reagent (from cell culture media).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeExosomes
Quantity6 mL
Sample TypeSerum
Shipping ConditionRoom Temperature
Product TypeTotal Exosome Isolation
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.
Frequently asked questions (FAQs)
What is the best way to store my exosomes?
For the short-term, exosomes can be stored at 4 degrees C for up to 1 week. For the long-term, exosomes can be stored at -20 degrees C or -80 degrees C. When storing exosomes for the long term, it is important to consider whether they will need to be thawed more than once for the target application. If multiple applications (and thus multiple thaws) will be used for analysis, then we recommend aliquoting the exosome resuspensions into multiple tubes so that each tube will only undergo one freeze/thaw cycle. We have found that multiple freeze thaw cycles can cause damage to the exosomes and reduce their numbers.
There are two protocol options for exosome isolation from plasma samples, which one should I choose?
Unlike serum, plasma contains numerous clotting factors and some additional proteins that can make it difficult to work with. We‘ve provided two protocol options, one with proteinase K (PK) and one without, in order to ease this difficulty. The protocol using PK is most useful when the end goal is analysis of the RNA or protein cargo contained inside the exosomes. It can also be used to isolate exosomes for use in other downstream applications, but it is most useful for RNA and protein analysis. The protocol without PK also isolates good quality exosomes, just not quite as pure as the PK protocol. The “no PK” protocol is more useful for isolating exosomes that will be used for surface protein analysis or electron microscopy identification.
My Westerns do not seem to work after exosome isolation. Can you help?
There are several possible reasons why Western blotting analysis is challenging:
1. Not enough sample volume added. Exosomes can contain a fairly low amount of protein cargo, so for an initial experiment we recommend adding as much of the sample as possible.
2. Antibody concentration should be titrated. Also, they should ideally be used fresh and need to be stored properly.
3. Depending on the exosomal surface marker, certain gel conditions might be more optimal for the target antibody (e.g., reducing/nonreducing and denaturing/nondenaturing). We suggest checking with the manufacturer and exosome community about which Western blotting conditions are recommended for the specific marker you are targeting and the specific antibody you are using.
4. General Western techniques. Westerns can be tricky so we recommend the use of a positive control for initial testing to make sure the entire workflow is functioning as it should. Any protein or antibody can be used as long as they meet the conditions you need (e.g., denaturing vs. non-denaturing). In addition, when picking the protein, try to steer clear of those that are present at very high or very low concentrations in your sample to prevent overloading the blot or total absence of signal.
How much RNA can be recovered from the exosomes?
This can vary depending on the sample type, volume of sample, isolation method, and exosome content/concentration. Listed below are some examples:
1) When exosomes are isolated from 30 mL of HeLa cell culture medium using the Total Exosome Isolation Reagent, it is possible to recover approximately 8 ng exosomal RNA.
2) For exosomes recovered from 4 mL serum, approximately 2 ng exosomal RNA can be obtained.
In both cases, these amounts of RNA are sufficient for RNA library prep for Ion PGM or Ion Proton sequencing. For real-time PCR analysis, substantially smaller amounts of RNA are needed and much lower sample volumes can be used. For example, RNA recovered from 3 µL serum or 30 µL medium is enough for one qRT-PCR reaction.
I'm using the Total Exosome RNA & Protein Isolation Kit. When ethanol is added to buffer 2/3, the solution turns turbid. Does this affect the efficiency of RNA recovery?
No, the described effect does not have a negative impact on the RNA recovery.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link