Invitrogen™

Total Exosome RNA & Protein Isolation Kit

Catalog number: 4478545
Have Questions?
Invitrogen™

Total Exosome RNA & Protein Isolation Kit

Catalog number: 4478545
Have Questions?
Catalog Number
4478545
Unit Size
40 preps
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
447854540 preps
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Total Exosome RNA and Protein Isolation Kit enables isolation of total RNA and proteins from exosome samples extracted from biological fluids (e.g., by using Total Exosome Isolation (from cell culture media) or Total Exosome Isolation (from serum)).
• Extracts highly pure total RNA (including small RNA fraction) from exosomes
• Recovers protein and RNA from the same sample
• Compatible with all protocols for exosome isolation

The Total Exosome RNA and Protein Isolation Kit is designed for the isolation of both RNA and protein from pre-isolated or enriched exosomes (not supplied). It is suitable for studies of RNA expression (specifically miRNA), processing, or function. The kit allows recovery of both RNA and proteins from the same purified exosome preparation. One portion of the sample is subjected to organic extraction, followed by immobilization of RNA on glass-fiber filters to purify total RNA or enrich for small RNA species. The other portion is then used directly for common protein applications such as Western blotting.

High yields of ultra-pure RNA can be prepared in about 30–60 minutes. The RNA can then be used in a variety of downstream applications, including qRT-PCR, high-throughput sequencing (e.g., using the Ion Torent™ PGM™, Ion Torrent™ Proton™, or SOLiD™ sequencing instruments), microarray analysis, solution hybridization assays, and blot hybridization.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Western Blot, microRNA Analysis
Product Type
Total Exosome Isolation
Final Product Type
Total RNA, Protein, micro RNA
High-throughput Compatibility
Not High-throughput Compatible (Manual)
Isolation Technology
Spin Column (Glass Fiber Filter), Organic Extraction
Purification Target
Total RNA, Protein, RNA & Protein, miRNA
Sample Type
Organelle & Vesicle-Related Samples, Exosomes
Shipping Condition
Room Temperature

Contents & Storage

Store at 2°C to 8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text