Thermo Scientific Chemicals

Sodium sulfate, 99%, for biochemistry, anhydrous

Catalog number: 447920025
2.5 kg, Each
Thermo Scientific Chemicals

Sodium sulfate, 99%, for biochemistry, anhydrous

Catalog number: 447920025
2.5 kg, Each
Quantity
Catalog number: 447920025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7757-82-6
IUPAC Name
disodium sulfate
Molecular Formula
Na2O4S
InChI Key
PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([O-])(=O)=O
Insoluble matter
=<0.01 %
pH
5.0 to 9.2
Loss on drying
=<0.5 %
Acidity/alkalinity
Passes test
Heavy metals (as Pb)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text