Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry

Catalog number: 447960025
2.5 kg, Each
Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, 98%, for biochemistry

Catalog number: 447960025
2.5 kg, Each
Quantity
Catalog number: 447960025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10101-89-0
IUPAC Name
trisodium dodecahydrate phosphate
Molecular Formula
H24Na3O16P
InChI Key
ASTWEMOBIXQPPV-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Calcium (Ca)
=<0.005 %
Magnesium (Mg)
=<0.0005 %
Appearance (Color)
Colorless to white
Appearance (Form)
Crystalline powder
Titration excess HCl
97.5 to 102.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text