Calcium sulfate dihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Calcium sulfate dihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium sulfate dihydrate, 98%, for biochemistry, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 448035000
Price (USD)/ Each
83.80
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
83.80
Add to cart
Calcium sulfate dihydrate, 98%, for biochemistry, Thermo Scientific Chemicals
Catalog number448035000
Price (USD)/ Each
83.80
-
Add to cart
Chemical Identifiers
IUPAC Namecalcium dihydrate sulfate
Molecular FormulaCaH4O6S
InChI KeyPASHVRUKOFIRIK-UHFFFAOYSA-L
SMILESO.O.[Ca++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)172.16
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Chloride (Cl)=<0.005 %
Iron (Fe)=<0.001 %
Magnesium (Mg)=<0.01 %
Potassium (K)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only