Chemical Identifiers
CAS56378-72-4
IUPAC Namepentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Molecular FormulaC4H12Mg5O19
InChI KeySXKPPTNZIUYFJR-UHFFFAOYSA-D
SMILESO.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more