Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium o
Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium o
Thermo Scientific Chemicals

Magnesium carbonate hydroxide pentahydrate, for biochemistry, 40.0-43.5% Magnesium o

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 448045000
Price (USD)/ Each
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS56378-72-4
IUPAC Namepentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Molecular FormulaC4H12Mg5O19
InChI KeySXKPPTNZIUYFJR-UHFFFAOYSA-D
SMILESO.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric40.0 to 43.5 % (MgO)
Acid insoluble substances=<0.05 % (in HCl)
Lead (Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only