Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals
Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Catalog number 448060025
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
Add to cart
Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals
Catalog number448060025
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS13472-36-1
IUPAC Nametetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular FormulaH20Na4O17P2
InChI KeyVZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILESO.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH9.5 to 10.5 (5% solution at 25.0°C)
Insoluble matter=<0.01 %
Sulfate (SO4)=<0.005 %
Nitrogen compounds (as N)=<0.001 %
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only