Thermo Scientific Chemicals

Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 448061000
100 g, Each
Thermo Scientific Chemicals

Sodium pyrophosphate decahydrate, 99-103%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 448061000
100 g, Each
Quantity
Catalog number: 448061000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13472-36-1
IUPAC Name
tetrasodium decahydrate (phosphonatooxy)phosphonate
Molecular Formula
H20Na4O17P2
InChI Key
VZWGHDYJGOMEKT-UHFFFAOYSA-J
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])([O-])=O
pH
9.5 to 10.5 (5% solution at 25.0°C)
Insoluble matter
=<0.01 %
Sulfate (SO4)
=<0.005 %
Nitrogen compounds (as N)
=<0.001 %
Appearance (Form)
Crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text