MicroAmp™ Optical 96-Well GPLE Reaction Plates with Barcode
Catalog number: 4481192
Have Questions?
Applied Biosystems™
MicroAmp™ Optical 96-Well GPLE Reaction Plates with Barcode
Catalog number: 4481192
Have Questions?
Catalog Number
4481192
Unit Size
10 plates
Price (USD)
107.00
Availability
-
Quantity
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
4481192
10 plates
107.00
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Applied Biosystems MicroAmp Optical 96-Well GPLE Reaction Plates with Barcode are validated and tested for optimal fit and performance on Applied Biosystems standard (0.2 mL) 96-well thermal cyclers, 0.2-mL real-time PCR instruments, and genetic analyzers.
Features of the MicroAmp Optical 96-Well GPLE Reaction Plates include: • Each lot is certified DNA-, RNase-, and PCR inhibitor-free • Labeled “For Laboratory Use” • Optimized for performance with Applied Biosystems instruments • Excellent heat transfer from thermal block to sample with thin-wall polypropylene wells • Raised deck for efficient plate sealing to help prevent evaporation and minimize contamination
MicroAmp Optical 96-well reaction plates are constructed from a single rigid piece of polypropylene and include a raised deck for efficient plate sealing. The thin-wall polypropylene wells provide excellent heat transfer from the thermal block to the sample for fast, efficient PCR amplification. These MicroAmp Optical 96-well reaction plates contain a single barcode (including a human readable barcode) on the south (row H1-H12) side of the plate.
With General Purpose Lab Equipment (GPLE) plastics, each lot of reaction plates is certified in an ISO 13485-registered facility to be free of DNA, RNase, and PCR inhibitors. GPLE plastics are ideal for use in diagnostic procedures.