Thermo Scientific Chemicals

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 448140050
5 kg, Each
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 448140050
5 kg, Each
Quantity
Catalog number: 448140050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7558-79-4
IUPAC Name
disodium hydrogen phosphate
Molecular Formula
HNa2O4P
InChI Key
BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES
[Na+].[Na+].OP([O-])([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Titration with HClO4
98.0 to 102.0 % (On dried substance)
Loss on drying
=<5 % (130°C)
pH
8.7 to 9.4 (5 % in water at 25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text